6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine

Lieferant: Apollo Scientific
Achtung

OR61524-250MG
APOSOR61524-250MGEA 33.5 EUR
APOSOR61524-250MG
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2-methyl-3H-imidazo[4,5-b]pyridine
Formel: C₇H₆BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00763284
CAS-Nummer: 42869-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR