7-Bromo-2-methyl-3H-imidazo[4,5-c]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-methyl-6-azabenzimidazole, 7-Bromo-2-methyl-1H-imidazo[4,5-c]pyridine

OR61530-250MG
APOSOR61530-250MGEA 217 EUR
APOSOR61530-250MG
7-Bromo-2-methyl-3H-imidazo[4,5-c]pyridine
7-Bromo-2-methyl-3H-imidazo[4,5-c]pyridine
Formel: C₇H₆BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18256908
CAS-Nummer: 929074-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR