6-Bromo-2,7-dimethyl-3H-imidazo[4,5-b]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2,4-dimethyl-7-azabenzimidazole
OR61531-250MG
APOSOR61531-250MGEA
174
EUR
APOSOR61531-250MG
6-Bromo-2,7-dimethyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2,7-dimethyl-3H-imidazo[4,5-b]pyridine
Formel:
C₈H₈BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08062977 CAS-Nummer: 954238-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...