6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-methyl-6-azabenzimidazole, 6-Chloro-2-methyl-1H-imidazo[4,5-c]pyridine
OR61532-1G
APOSOR61532-1GEA
989
EUR
APOSOR61532-1G
6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine
6-Chloro-2-methyl-3H-imidazo[4,5-c]pyridine
Formel:
C₇H₆ClN₃ Molecular Weight: 167,596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18256899 CAS-Nummer: 7205-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...