4-Bromo-2-methoxybenzenesulphonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

OR61543-1G
APOSOR61543-1GEA 394 EUR
APOSOR61543-1G
4-Bromo-2-methoxybenzenesulphonyl chloride 97%
4-Bromo-2-methoxybenzenesulphonyl chloride
Formel: C₇H₆BrClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD11847575
CAS-Nummer: 145915-29-3
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR