2-Bromo-3-chloro-5-iodobenzyl bromide ≥94%

Lieferant: Apollo Scientific

OR61548-250MG OR61548-1G
APOSOR61548-250MGEA 190 EUR
APOSOR61548-250MG APOSOR61548-1G
2-Bromo-3-chloro-5-iodobenzyl bromide ≥94%
2-Bromo-3-chloro-5-iodobenzyl bromide
Formel: C₇H₄Br₂ClI
Molecular Weight: 410.27212 g/mol
CAS-Nummer: 2092415-39-7
UN: 3261

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR