2-Bromo-3-chloro-5-iodobenzyl bromide ≥94%
Lieferant: Apollo Scientific
OR61548-250MG
OR61548-1G
APOSOR61548-250MGEA
190
EUR
APOSOR61548-250MG
APOSOR61548-1G
2-Bromo-3-chloro-5-iodobenzyl bromide ≥94%
2-Bromo-3-chloro-5-iodobenzyl bromide
Formel:
C₇H₄Br₂ClI Molecular Weight: 410.27212 g/mol |
CAS-Nummer:
2092415-39-7 UN: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...