(R)-2-Amino-4,4-dimethylpentanoic acid 97%

Lieferant: Apollo Scientific

Synonyme: H-D-4-Me-Leu-OH, H-β-tBu-D-Ala-OH

OR61606-1G
APOSOR61606-1GEA 172 EUR
APOSOR61606-1G
(R)-2-Amino-4,4-dimethylpentanoic acid 97%
(R)-2-Amino-4,4-dimethylpentanoic acid
Formel: C₇H₁₅NO₂
Molecular Weight: 145,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038404
CAS-Nummer: 88319-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR