1H-Pyrrol-3-carbaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR6181-250MG
OR6181-10G
OR6181-1G
APOSOR6181-250MGEA
21.9
EUR
APOSOR6181-250MG
APOSOR6181-10G
APOSOR6181-1G
1H-Pyrrol-3-carbaldehyd 95%
1H-Pyrrole-3-carbaldehyde
Formel:
C₅H₅NO Schmelzpunkt: 65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426733 CAS-Nummer: 7126-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...