5-Methyl-1H-pyrazol-4-carbaldehyd 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyl-3-methylpyrazole, 3-Methylpyrazole-4-carboxaldehyde
OR6183-5G
APOSOR6183-5GEA
58
EUR
APOSOR6183-5G
5-Methyl-1H-pyrazol-4-carbaldehyd 95%
3-Methyl-1H-pyrazole-4-carbaldehyde
Formel:
C5H6N2O Molecular Weight: 110.12 g/mol Schmelzpunkt: 105…110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01313822 CAS-Nummer: 112758-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...