1-[4-(Bromomethyl)phenyl]methanamine

Lieferant: Apollo Scientific

OR6190-1G
APOSOR6190-1GEA 415 EUR
APOSOR6190-1G
1-[4-(Bromomethyl)phenyl]methanamine
1-[4-(Bromomethyl)phenyl]methanamine
Formel: C₈H₁₀BrN
Lagertemperatur: Raumtemperatur
MDL: MFCD03410966
CAS-Nummer: 769057-30-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR