3-(Aminomethyl)benzoesäure Hydrochlorid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Aminomethyl)benzoic acid, HCl
OR6195-5G
OR6195-25G
APOSOR6195-5GEA
55.1
EUR
APOSOR6195-5G
APOSOR6195-25G
3-(Aminomethyl)benzoesäure Hydrochlorid 95%
3-(Aminomethyl)benzoesäure Hydrochlorid
Formel:
C₈H₁₀ClNO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD03791117 CAS-Nummer: 876-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...