Methyl-5-brom-1H-indazol-3-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-bromo-1H-indazole-3-carboxylate
OR62000-250MG
APOSOR62000-250MGEA
21.9
EUR
APOSOR62000-250MG
Methyl-5-brom-1H-indazol-3-carboxylat
Methyl-5-brom-1H-indazol-3-carboxylat
Formel:
C₉H₇BrN₂O₂ Molecular Weight: 255,0700 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07371572 CAS-Nummer: 78155-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...