Methyl-6-brom-1H-indazol-3-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-bromo-1H-indazole-3-carboxylate
OR62001-250MG
APOSOR62001-250MGEA
21.9
EUR
APOSOR62001-250MG
Methyl-6-brom-1H-indazol-3-carboxylat
Methyl-6-brom-1H-indazol-3-carboxylat
Formel:
C₉H₇BrN₂O₂ Molecular Weight: 255,0700 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07371573 CAS-Nummer: 885278-42-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...