(3S)-3-(Isopropylamino)piperidine, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: (3S)-3-[(Prop-2-yl)amino]piperidine, N1-BOC protected, tert-Butyl (3S)-3-[(prop-2-yl)amino]piperidine-1-carboxylate, (3S)-1-(tert-Butoxycarbonyl)-3-[(prop-2-yl)amino]piperidine

OR62007-1G
APOSOR62007-1GEA 626 EUR
APOSOR62007-1G
(3S)-3-(Isopropylamino)piperidine, N1-BOC protected
(3S)-3-(Isopropylamino)piperidine, N1-BOC protected
Formel: C₁₃H₂₆N₂O₂
Molecular Weight: 242,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19381682
CAS-Nummer: 1163286-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR