(3S)-3-(Isobutylamino)piperidine, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: (3S)-1-(tert-Butoxycarbonyl)-3-[(2-methylprop-1-yl)amino]piperidine, tert-Butyl (3S)-3-[(2-methylprop-1-yl)amino]piperidine-1-carboxylate, (3S)-3-[(2-Methylprop-1-yl)amino]piperidine, N1-BOC protected

OR62009-1G
APOSOR62009-1GEA 626 EUR
APOSOR62009-1G
(3S)-3-(Isobutylamino)piperidine, N1-BOC protected
(3S)-3-(Isobutylamino)piperidine, N1-BOC protected
Formel: C₁₄H₂₈N₂O₂
Molecular Weight: 256,384 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19381684
CAS-Nummer: 1027346-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR