Acetobromo-α-D-glucuronic acid methyl ester 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

OR6200T-1G OR6200T-5G OR6200T-25G
APOSOR6200T-1GEA 50.9 EUR
APOSOR6200T-1G APOSOR6200T-5G APOSOR6200T-25G
Acetobromo-α-D-glucuronic acid methyl ester 95+%
Acetobromo-alpha-D-glucuronic acid methyl ester
Formel: C₁₃H₁₇BrO₉
Molecular Weight: 397.174 g/mol
Schmelzpunkt: 82…110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00061613
CAS-Nummer: 21085-72-3
EINECS: 244-203-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR