Acetobromo-α-D-glucuronic acid methyl ester 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
OR6200T-1G
OR6200T-5G
OR6200T-25G
APOSOR6200T-1GEA
50.9
EUR
APOSOR6200T-1G
APOSOR6200T-5G
APOSOR6200T-25G
Acetobromo-α-D-glucuronic acid methyl ester 95+%
Acetobromo-alpha-D-glucuronic acid methyl ester
Formel:
C₁₃H₁₇BrO₉ Molecular Weight: 397.174 g/mol Schmelzpunkt: 82…110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00061613 CAS-Nummer: 21085-72-3 EINECS: 244-203-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...