(3S)-3-{[(Pyridin-2-yl)methyl]amino}piperidine, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: 2-({[(3S)-1-(tert-Butoxycarbonyl)piperidin-3-yl]amino}methyl)pyridine, (3S)-1-(tert-Butoxycarbonyl)-3-{[(pyridin-2-yl)methyl]amino}piperidine, tert-Butyl (3S)-3-{[(pyridin-2-yl)methyl]amino}piperidine-1-carboxylate

OR62011-1G
APOSOR62011-1GEA 626 EUR
APOSOR62011-1G
(3S)-3-{[(Pyridin-2-yl)methyl]amino}piperidine, N1-BOC protected
(3S)-3-{[(Pyridin-2-yl)methyl]amino}piperidine, N1-BOC protected
Formel: C₁₆H₂₅N₃O₂
Molecular Weight: 291,389 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19381686
CAS-Nummer: 1349702-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR