10-Phenyldecylphosphonic acid 97%
Lieferant: Apollo Scientific
OR62068-1G
APOSOR62068-1GEA
567
EUR
APOSOR62068-1G
10-Phenyldecylphosphonic acid 97%
10-Phenyldecylphosphonic acid
Formel:
C₁₆H₂₇O₃P Molecular Weight: 298,358 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32665672 CAS-Nummer: 1429741-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...