5-Ethynyl-1H-pyrazolo[4,3-b]pyridine
Lieferant: Apollo Scientific
Synonyme:
(1H-Pyrazolo[4,3-b]pyridin-5-yl)acetylene, 5-Ethynyl-4-azaindazole
OR62078-250MG
APOSOR62078-250MGEA
746
EUR
APOSOR62078-250MG
5-Ethynyl-1H-pyrazolo[4,3-b]pyridine
5-Ethynyl-1H-pyrazolo[4,3-b]pyridine
Formel:
C₈H₅N₃ Molecular Weight: 143,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22380512 CAS-Nummer: 1374652-56-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...