N-Methyl-N-[1H-indol-3-yl)methyl]amine

Lieferant: Apollo Scientific

OR6211-1G
APOSOR6211-1GEA 654 EUR
APOSOR6211-1G
N-Methyl-N-[1H-indol-3-yl)methyl]amine
N-Methyl-N-[1H-indol-3-yl)methyl]amine
Formel: C₁₀H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03011586
CAS-Nummer: 36284-95-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR