7-Nitroindol

Lieferant: Apollo Scientific
Achtung

OR6214-25G OR6214-5G
APOSOR6214-25GEA 81.2 EUR
APOSOR6214-25G APOSOR6214-5G
7-Nitroindol
7-Nitroindol
Formel: C₈H₆N₂O₂
Molecular Weight: 162.15 g/mol
Schmelzpunkt: 96…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005683
CAS-Nummer: 6960-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR