8-Chloro-6-iodo-2H-benzo[d][1,3]oxazine-2,4(1H)-dione ≥94%

Lieferant: Apollo Scientific

OR62141-250MG OR62141-1G
APOSOR62141-250MGEA 232 EUR
APOSOR62141-250MG APOSOR62141-1G
8-Chloro-6-iodo-2H-benzo[d][1,3]oxazine-2,4(1H)-dione ≥94%
8-Chloro-6-iodo-2H-benzo[d][1,3]oxazine-2,4(1H)-dione
Formel: C₈H₃ClINO₃
Molecular Weight: 323.47179 g/mol
CAS-Nummer: 2609110-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR