2-Brom-3-hydroxypyridin ≥95%

Lieferant: Apollo Scientific

OR62149-25G OR62149-100G
APOSOR62149-25GEA 21.9 EUR
APOSOR62149-25G APOSOR62149-100G
2-Brom-3-hydroxypyridin ≥95%
2-Brom-3-hydroxypyridin
Formel: C₅H₄BrNO
MDL: MFCD00006220
CAS-Nummer: 6602-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR