3-(2-Ethoxyphenoxy)-1,2-propanediol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR62179-1G
APOSOR62179-1GEA
719
EUR
APOSOR62179-1G
3-(2-Ethoxyphenoxy)-1,2-propanediol 95%
3-(2-Ethoxyphenoxy)-1,2-propanediol
Formel:
C₁₁H₁₆O₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD00867665 CAS-Nummer: 63834-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...