Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%

Lieferant: Apollo Scientific
Achtung

OR62216-1G
APOSOR62216-1GEA 574 EUR
APOSOR62216-1G
Methyl 4-bromo-2-iodo-5-methoxybenzoate 95%
Methyl 4-bromo-2-iodo-5-methoxybenzoate
Formel: C₉H₈BrIO₃
CAS-Nummer: 2384857-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR