3-(1H-Pyrazol-4-yl)benzoesäure 95%

Lieferant: Apollo Scientific
Achtung

OR6231-1G OR6231-250MG OR6231-5G OR6231-500MG OR6231-2.5G
APOSOR6231-1GEA 112 EUR
APOSOR6231-1G APOSOR6231-250MG APOSOR6231-5G APOSOR6231-500MG APOSOR6231-2.5G
3-(1H-Pyrazol-4-yl)benzoesäure 95%
3-(1H-Pyrazol-4-yl)benzoic acid
Formel: C₁₀H₈N₂O₂
Lagertemperatur: Gefrierschrank
MDL: MFCD06739075
CAS-Nummer: 1002535-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR