4-Pyrimidin-2-ylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Pyrimidin-2-yl-benzoic acid

OR6232-100MG
APOSOR6232-100MGEA 26.1 EUR
APOSOR6232-100MG
4-Pyrimidin-2-ylbenzoic acid 95%
4-Pyrimidin-2-ylbenzoic acid
Formel: C₁₁H₈N₂O₂
Molecular Weight: 200,1900 g/mol
Schmelzpunkt: 238…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318668
CAS-Nummer: 199678-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR