3-Pyrimidin-2-ylbenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR6233-1G OR6233-5G
APOSOR6233-1GEA 168 EUR
APOSOR6233-1G APOSOR6233-5G
3-Pyrimidin-2-ylbenzoic acid 95%
3-Pyrimidin-2-ylbenzoic acid
Formel: C₁₁H₈N₂O₂
Schmelzpunkt: 180…182 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 579476-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR