2-Pyrid-4-ylbenzoesäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR6234-1G
OR6234-2.5G
APOSOR6234-1GEA
249
EUR
APOSOR6234-1G
APOSOR6234-2.5G
2-Pyrid-4-ylbenzoesäure 95%
2-Pyrid-4-ylbenzoesäure
Formel:
C₁₂H₉NO₂ Schmelzpunkt: 180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056297 CAS-Nummer: 133362-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...