3-(1H-Imidazol-4-yl)benzoic acid
Lieferant: Apollo Scientific
OR6240-1G
APOSOR6240-1GEA
673
EUR
APOSOR6240-1G
3-(1H-Imidazol-4-yl)benzoic acid
3-(1H-Imidazol-4-yl)benzoic acid
Formel:
C₁₀H₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08056299 CAS-Nummer: 912569-71-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...