4-Amino-3-brombenzoesäure 98%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-3-bromobenzoic Acid

OR6241-25G
APOSOR6241-25GEA 58 EUR
APOSOR6241-25G
4-Amino-3-brombenzoesäure 98%
4-Amino-3-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,0320 g/mol
Schmelzpunkt: 211…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407439
CAS-Nummer: 6311-37-1
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR