4-Piperdino-2-pyridine carboxylic acid

Lieferant: Apollo Scientific

OR6249-1G
APOSOR6249-1GEA 386 EUR
APOSOR6249-1G
4-Piperdino-2-pyridine carboxylic acid
4-Piperdino-2-pyridine carboxylic acid
Formel: C₁₁H₁₄N₂O₂
Molecular Weight: 206,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06382852
CAS-Nummer: 500356-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR