2-Hydrazinoisonicotinic acid

Lieferant: Apollo Scientific
Achtung

OR6257-1G OR6257-250MG OR6257-500MG
APOSOR6257-1GEA 458 EUR
APOSOR6257-1G APOSOR6257-250MG APOSOR6257-500MG
2-Hydrazinoisonicotinic acid
2-Hydrazinoisonicotinic acid
Formel: C₆H₇N₃O₂
Schmelzpunkt: 232 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07786611
CAS-Nummer: 887589-25-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR