5-Brom-2-chlornicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR6261-25G
APOSOR6261-25GEA
29
EUR
APOSOR6261-25G
5-Brom-2-chlornicotinsäure 97%
5-Brom-2-chlornicotinsäure
Formel:
C₆H₃BrClNO₂ Schmelzpunkt: 172…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844847 CAS-Nummer: 29241-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...