3-Pyridinylessigsäurehydrochlorid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

OR6262-25G OR6262-100G
APOSOR6262-25GEA 21.9 EUR
APOSOR6262-25G APOSOR6262-100G
3-Pyridinylessigsäurehydrochlorid 95%
2-(Pyridin-3-yl)acetic acid hydrochloride
Formel: C₇H₈ClNO₂
Schmelzpunkt: 161…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012819
CAS-Nummer: 6419-36-9
EINECS: 229-148-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR