5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific

OR6268-1G
APOSOR6268-1GEA 0 EUR
APOSOR6268-1G
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid
5-(1-Methylimidazol-2-yl)thiophene-2-carboxylic acid
Formel: C₉H₈N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08056306
CAS-Nummer: 914637-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR