4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
Lieferant: Apollo Scientific
Synonyme:
4-Methyl-2-(pyridin-3-yl)thiazole-5-carboxylic acid
OR6274-1G
APOSOR6274-1GEA
91.5
EUR
APOSOR6274-1G
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
4-Methyl-2-(3-pyridinyl)-5-thiazolcarbonsäure
Formel:
C₁₀H₈N₂O₂S Molecular Weight: 220,25 g/mol Schmelzpunkt: 244…248 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00171753 CAS-Nummer: 39091-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...