5-Methyloxazole-4-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxazolecarboxylicacid, 5-methyl-, 5-Methyl-1,3-oxazole-4-carboxylic acid, 4-Carboxy-5-methyl-1,3-oxazole

OR6279-1G
APOSOR6279-1GEA 21.9 EUR
APOSOR6279-1G
5-Methyloxazole-4-carboxylic acid 98%
5-Methyloxazole-4-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056309
CAS-Nummer: 103879-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR