N-Palmitoyl ceramide

Lieferant: Apollo Scientific

Synonyme: D-erythro-Sphingosine, N-Palmitoyl-, N-Palmitoyl-D-erythro-sphingosine, C16 Ceramide, N-Hexadecanoyl-D-erythro-sphingosine

OR6290T-25MG OR6290T-5MG OR6290T-50MG
APOSOR6290T-25MGEA 319 EUR
APOSOR6290T-25MG APOSOR6290T-5MG APOSOR6290T-50MG
N-Palmitoyl ceramide
N-Palmitoyl ceramide

A predominant, rather hydrophobic natural ceramide. Activates a cytosolic serine/threonine protein phosphate & stimulates ceramide-activated protein kinase.

Formel: C₃₄H₆₇NO₃
Schmelzpunkt: 94…95 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00871748
CAS-Nummer: 24696-26-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR