3-Chlor-2-nitroanilin 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR63031-5G
APOSOR63031-5GEA
58
EUR
APOSOR63031-5G
3-Chlor-2-nitroanilin 97%
3-Chlor-2-nitroanilin
Formel:
C₆H₅ClN₂O₂ Schmelzpunkt: 106…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01317803 CAS-Nummer: 59483-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...