2-Mercapto-1H-benzimidazole-5-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2-Mercapto-5-benzimidazolecarboxylic Acid, 2-Thio-1H-benzimidazole-5-carboxylic acid

OR6308-5G
APOSOR6308-5GEA 120 EUR
APOSOR6308-5G
2-Mercapto-1H-benzimidazole-5-carboxylic acid 97%
2-Mercapto-1H-benzimidazole-5-carboxylic acid
Formel: C₈H₆N₂O₂S
Molecular Weight: 194,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318607
CAS-Nummer: 58089-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR