5-Oxo-1-pyridin-3-ylpyrrolidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR6336-250MG
APOSOR6336-250MGEA 275 EUR
APOSOR6336-250MG
5-Oxo-1-pyridin-3-ylpyrrolidine-3-carboxylic acid 95%
5-Oxo-1-pyridin-3-ylpyrrolidine-3-carboxylic acid
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,2 g/mol
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07364771
CAS-Nummer: 914637-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR