Imidazo[1,2-a]pyrimidine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR6351-1G OR6351-250MG
APOSOR6351-1GEA 165 EUR
APOSOR6351-1G APOSOR6351-250MG
Imidazo[1,2-a]pyrimidine-2-carboxylic acid 97%
Imidazo[1,2-a]pyrimidine-2-carboxylic acid
Formel: C₇H₅N₃O₂
Schmelzpunkt: 238…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03419484
CAS-Nummer: 64951-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR