5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR6352-100MG
APOSOR6352-100MGEA 121 EUR
APOSOR6352-100MG
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid 95%
5H-Imidazo[1,2-b]pyrazole-2-carboxylic acid
Formel: C₆H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056315
CAS-Nummer: 914637-58-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR