Boc-L-norvaline 98%

Lieferant: Apollo Scientific

Synonyme: Boc-Nva-OH, N-(tert-Butoxycarbonyl)-L-norvaline, (2S)-2-Aminopentanoic acid, N-BOC protected, N-Boc-L-norvaline

OR6368-1G OR6368-5G
APOSOR6368-1GEA 72.5 EUR
APOSOR6368-1G APOSOR6368-5G
Boc-L-norvaline 98%
Boc-L-norvaline
Formel: C₁₀H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00037268
CAS-Nummer: 53308-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR