D,L-Anatabine 94%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,2,3,6-Tetrahydro-2,3'-bipyridine, (R,S)-Anatabine

OR6370T-100MG
APOSOR6370T-100MGEA 72.5 EUR
APOSOR6370T-100MG
D,L-Anatabine 94%
D,L-Anatabine

A minor component in tobacco.

Formel: C₁₀H₁₂N₂
Molecular Weight: 160.22 g/mol
Siedepunkt: 116…117 °C/1mmHg
Dichte: 1.086 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00078278
CAS-Nummer: 2743-90-0
UN: 2810
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR