4-Oxocyclohexanecarbonyl chloride

Lieferant: Apollo Scientific

OR6382-1G OR6382-250MG OR6382-500MG
APOSOR6382-1GEA 0 EUR
APOSOR6382-1G APOSOR6382-250MG APOSOR6382-500MG
4-Oxocyclohexanecarbonyl chloride
4-Oxocyclohexanecarbonyl chloride
Formel: C₇H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08056325
CAS-Nummer: 914637-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR