4-(Pyrimidin-2-yl)bromobenzene

Lieferant: Apollo Scientific

Synonyme: 2-(4-Bromophenyl)pyrimidine

OR6403-1G
APOSOR6403-1GEA 255 EUR
APOSOR6403-1G
4-(Pyrimidin-2-yl)bromobenzene
4-(Pyrimidin-2-yl)bromobenzene
Formel: C₁₀H₇BrN₂
Molecular Weight: 235,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056329
CAS-Nummer: 210354-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR